Assay Details
Target Gene Details
Entrez Gene ID: | 9971 |
Gene Name: | nuclear receptor subfamily 1 group H member 4 |
Gene Aliases: |
BAR, FXR, HRR-1, HRR1, RIP14 |
Location: |
Chr.12:100473773-100563867 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR1H4 | NM_001206977.1 | NP_001193906.1 | ||
| NM_001206978.1 | NP_001193907.1 | |||
| NM_001206979.1 | NP_001193908.1 | |||
| NM_005123.3 | NP_005114.1 | |||
| NR_135146.1 | ||||
| XM_011539040.2 | XP_011537342.1 | |||
| AB307699.1 | ||||
| AF384555.1 | AAK60271.1 | |||
| AK312959.1 | ||||
| BC130573.1 | ||||
| BC144184.1 | ||||
| HQ709175.1 | ||||
| U68233.1 | AAB08107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559995 | Chr.12:100487230 - 100542549 on Build GRCh38 | Loss |
|
| nsv559964 | Chr.12:99872025 - 100538490 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76175497] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map