Assay Details
Target Gene Details
Entrez Gene ID: | 8825 |
Gene Name: | lin-7 homolog A, crumbs cell polarity complex component |
Gene Aliases: |
LIN-7A, LIN7, MALS-1, TIP-33, VELI1 |
Location: |
Chr.12:80792520-80937915 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIN7A | NM_001324423.1 | NP_001311352.1 | ||
| NM_004664.3 | NP_004655.1 | |||
| NR_136887.1 | ||||
| NR_136888.1 | ||||
| XM_011538928.2 | XP_011537230.1 | |||
| AF087693.1 | AAC78481.1 | |||
| AF173081.1 | AAD48500.1 | |||
| AK309256.1 | ||||
| AK315321.1 | ||||
| BC099921.1 | ||||
| BC118609.1 | ||||
| BC122561.1 | ||||
| CR407680.1 | CAG28608.1 |
Target Gene Details
Entrez Gene ID: | 105369869 |
Gene Name: | uncharacterized LOC105369869 |
Gene Aliases: |
- |
Location: |
Chr.12:80933598-80937632 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369869 | XM_011539062.2 | 2 | 3446 | XP_011537364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv789 | Chr.12:80920377 - 80966429 on Build GRCh38 | Deletion |
|
| esv3580346 | Chr.12:80933865 - 80939144 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map