Assay Details
Target Gene Details
Entrez Gene ID: | 117177 |
Gene Name: | RAB3A interacting protein |
Gene Aliases: |
RABIN3, RABIN8 |
Location: |
Chr.12:69738007-69823204 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3IP | NM_022456.4 | NP_071901.2 | ||
| NM_175623.3 | NP_783322.1 | |||
| NM_175624.3 | NP_783323.1 | |||
| NM_175625.3 | NP_783324.1 | |||
| NR_103519.1 | ||||
| NR_103520.1 | ||||
| XM_006719226.3 | XP_006719289.3 | |||
| XM_006719227.3 | XP_006719290.3 | |||
| XM_017018774.1 | XP_016874263.1 | |||
| XM_017018775.1 | XP_016874264.1 | |||
| XM_017018776.1 | XP_016874265.1 | |||
| XM_017018777.1 | XP_016874266.1 | |||
| AJ312896.1 | CAC59835.1 | |||
| AJ312897.1 | CAC59836.1 | |||
| AJ312898.1 | CAC59837.1 | |||
| AJ312899.1 | CAC59838.1 | |||
| AJ312900.1 | CAC59839.1 | |||
| AJ312901.1 | CAC59840.1 | |||
| AK026201.1 | BAB15391.1 | |||
| AK057844.1 | ||||
| AK225441.1 | ||||
| AK314547.1 | ||||
| BC059358.1 | AAH59358.1 | |||
| DA211792.1 | ||||
| DA874650.1 | ||||
| DB460944.1 |
More Information
Set Membership: |
|

Genomic Map