Assay Details
Target Gene Details
Entrez Gene ID: | 84790 |
Gene Name: | tubulin alpha 1c |
Gene Aliases: |
TUBA6, bcm948 |
Location: |
Chr.12:49227926-49273338 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBA1C | NM_001303114.1 | NP_001290043.1 | ||
| NM_001303115.1 | NP_001290044.1 | |||
| NM_001303116.1 | NP_001290045.1 | |||
| NM_001303117.1 | NP_001290046.1 | |||
| AK293589.1 | ||||
| AL539200.3 | ||||
| DA686435.1 | ||||
| DC366449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518041 | Chr.12:49224260 - 49251457 on Build GRCh38 | Loss |
|
| nsv701 | Chr.12:49226851 - 49251249 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117888104,rs73306242] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map