Assay Details
Target Gene Details
Entrez Gene ID: | 51699 |
Gene Name: | VPS29, retromer complex component |
Gene Aliases: |
DC15, DC7, PEP11 |
Location: |
Chr.12:110491523-110502140 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Set Membership: |
|

Genomic Map