Assay Details
Target Gene Details
Entrez Gene ID: | 54434 |
Gene Name: | slingshot protein phosphatase 1 |
Gene Aliases: |
SSH1L |
Location: |
Chr.12:108782690-108857606 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSH1 | NM_001161330.1 | 14 | 2246 | NP_001154802.1 |
| NM_001161331.1 | 13 | 2338 | NP_001154803.1 | |
| NM_018984.3 | NP_061857.3 | |||
| XM_005268984.4 | XP_005269041.1 | |||
| XM_005268985.2 | XP_005269042.1 | |||
| XM_011538497.1 | XP_011536799.1 | |||
| XM_011538499.1 | XP_011536801.1 | |||
| XM_011538500.2 | XP_011536802.1 | |||
| XM_011538501.2 | XP_011536803.1 | |||
| XM_017019491.1 | XP_016874980.1 | |||
| XM_017019492.1 | XP_016874981.1 | |||
| XM_017019493.1 | XP_016874982.1 | |||
| XM_017019494.1 | XP_016874983.1 | |||
| AB037719.1 | BAA92536.1 | |||
| AB072355.1 | BAB84114.1 | |||
| AB072356.1 | 14 | 2159 | BAB84115.1 | |
| AK095421.1 | 13 | 2339 | BAC04546.1 | |
| BC062341.1 | AAH62341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763002 | Chr.12:108780333 - 108811466 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map