Assay Details
Target Gene Details
Entrez Gene ID: | 8550 |
Gene Name: | mitogen-activated protein kinase-activated protein kinase 5 |
Gene Aliases: |
MAPKAP-K5, MK-5, MK5, PRAK |
Location: |
Chr.12:111842228-111902225 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044606 | Chr.12:111885161 - 111989037 on Build GRCh38 | Gain |
|
| nsv1045488 | Chr.12:111883935 - 111922897 on Build GRCh38 | Gain |
|
| esv1010936 | Chr.12:111890837 - 111902032 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs184956284] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map