Assay Details
Target Gene Details
Entrez Gene ID: | 5463 |
Gene Name: | POU class 6 homeobox 1 |
Gene Aliases: |
BRN5, MPOU, TCFB1 |
Location: |
Chr.12:51186936-51218134 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79809537] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map