Assay Details
Target Gene Details
Entrez Gene ID: | 3856 |
Gene Name: | keratin 8 |
Gene Aliases: |
CARD2, CK-8, CK8, CYK8, K2C8, K8, KO |
Location: |
Chr.12:52897187-52949866 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT8 | NM_001256282.1 | NP_001243211.1 | ||
| NM_001256293.1 | NP_001243222.1 | |||
| NR_045962.1 | ||||
| AK290938.1 | ||||
| AK310257.1 | ||||
| AK315826.1 | ||||
| DA051933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832416 | Chr.12:52779656 - 52971582 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75823692] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map