Assay Details
Target Gene Details
Entrez Gene ID: | 636 |
Gene Name: | BICD cargo adaptor 1 |
Gene Aliases: |
BICD |
Location: |
Chr.12:32106736-32383637 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BICD1 | NM_001003398.1 | NP_001003398.1 | ||
| NM_001714.2 | NP_001705.2 | |||
| XM_005253465.3 | XP_005253522.1 | |||
| XM_005253466.2 | XP_005253523.1 | |||
| XM_006719134.3 | XP_006719197.1 | |||
| XM_011520812.2 | XP_011519114.1 | |||
| XM_011520813.1 | XP_011519115.1 | |||
| XM_011520814.2 | XP_011519116.1 | |||
| XM_011520815.2 | XP_011519117.1 | |||
| XM_011520816.2 | XP_011519118.1 | |||
| XM_017019797.1 | XP_016875286.1 | |||
| AK290188.1 | ||||
| AK313430.1 | ||||
| BC136372.1 | ||||
| U90028.1 | AAB94805.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041733 | Chr.12:32094493 - 32159613 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map