Assay Details
Target Gene Details
Entrez Gene ID: | 7846 |
Gene Name: | tubulin alpha 1a |
Gene Aliases: |
B-ALPHA-1, LIS3, TUBA3 |
Location: |
Chr.12:49184795-49189324 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBA1A | NM_001270399.1 | NP_001257328.1 | ||
| NM_001270400.1 | NP_001257329.1 | |||
| NM_006009.3 | NP_006000.2 | |||
| AF141347.1 | AAD33871.1 | |||
| AK057842.1 | ||||
| AK289483.1 | ||||
| AK297775.1 | ||||
| AK298838.1 | ||||
| AU136397.2 | ||||
| BC006468.1 | AAH06468.1 | |||
| BC050637.2 | AAH50637.1 | |||
| DC340080.1 |
More Information
Set Membership: |
|

Genomic Map