Assay Details
Target Gene Details
Entrez Gene ID: | 341350 |
Gene Name: | ovochymase 1 |
Gene Aliases: |
OVCH |
Location: |
Chr.12:29426871-29499693 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OVCH1 | NM_183378.2 | NP_899234.2 | ||
| XM_011520638.2 | XP_011518940.1 | |||
| XM_011520639.2 | XP_011518941.1 | |||
| XM_011520640.2 | 20 | 4547 | XP_011518942.1 | |
| XM_011520641.2 | XP_011518943.1 | |||
| XM_017019256.1 | XP_016874745.1 | |||
| XM_017019257.1 | XP_016874746.1 |
Target Gene Details
Entrez Gene ID: | 101055625 |
Gene Name: | OVCH1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:29389294-29487324 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OVCH1-AS1 | NR_073170.1 | |||
| NR_073171.1 | ||||
| NR_073172.1 | ||||
| AA910547.1 | ||||
| BQ058205.1 | ||||
| BX119718.1 | ||||
| CA307018.1 | ||||
| HY017985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580214 | Chr.12:29448596 - 29465340 on Build GRCh38 | Loss |
|
| nsv469190 | Chr.12:29459492 - 29658538 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map