Assay Details
Target Gene Details
Entrez Gene ID: | 8086 |
Gene Name: | aladin WD repeat nucleoporin |
Gene Aliases: |
AAA, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003 |
Location: |
Chr.12:53307456-53321628 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAAS | NM_001173466.1 | NP_001166937.1 | ||
| NM_015665.5 | NP_056480.1 | |||
| XM_011538778.1 | XP_011537080.1 | |||
| XM_011538780.1 | XP_011537082.1 | |||
| AF226048.1 | AAF86948.1 | |||
| AJ289857.1 | CAC19017.1 | |||
| AK000833.1 | BAA91394.1 | |||
| AK094302.1 | ||||
| AK222509.1 | BAD96229.1 | |||
| AK293342.1 | ||||
| AY237818.1 | AAP69911.1 | |||
| BC000659.2 | AAH00659.1 | |||
| BT006912.1 | AAP35558.1 | |||
| DC393722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map