Assay Details
Target Gene Details
Entrez Gene ID: | 101928002 |
Gene Name: | uncharacterized LOC101928002 |
Gene Aliases: |
- |
Location: |
Chr.12:69713633-69738568 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928002 | NR_110072.1 | 1 | 93 |
Target Gene Details
Entrez Gene ID: | 117177 |
Gene Name: | RAB3A interacting protein |
Gene Aliases: |
RABIN3, RABIN8 |
Location: |
Chr.12:69738007-69823204 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3IP | XM_006719226.3 | XP_006719289.3 | ||
| XM_006719227.3 | XP_006719290.3 | |||
| XM_017018775.1 | XP_016874264.1 |
More Information
Set Membership: |
|

Genomic Map