Assay Details
Target Gene Details
Entrez Gene ID: | 160777 |
Gene Name: | coiled-coil domain containing 60 |
Gene Aliases: |
- |
Location: |
Chr.12:119334712-119541047 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC60 | NM_178499.4 | NP_848594.2 | ||
| XM_017018911.1 | XP_016874400.1 | |||
| XM_017018912.1 | XP_016874401.1 | |||
| XM_017018913.1 | XP_016874402.1 | |||
| AK310830.1 | ||||
| BC040553.1 | AAH40553.1 | |||
| DB204596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832528 | Chr.12:119200406 - 119355596 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111438396] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map