Assay Details
Target Gene Details
Entrez Gene ID: | 8550 |
Gene Name: | mitogen-activated protein kinase-activated protein kinase 5 |
Gene Aliases: |
MAPKAP-K5, MK-5, MK5, PRAK |
Location: |
Chr.12:111842228-111902225 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPKAPK5 | XM_017020127.1 | 15 | 3483 | XP_016875616.1 |
| XM_017020128.1 | 15 | 1934 | XP_016875617.1 | |
| XM_017020132.1 | 16 | 3531 | XP_016875621.1 | |
| XM_017020133.1 | 15 | 3303 | XP_016875622.1 | |
| XM_017020134.1 | 15 | 3129 | XP_016875623.1 | |
| XM_017020135.1 | 14 | 3212 | XP_016875624.1 | |
| XM_017020136.1 | 15 | 3297 | XP_016875625.1 | |
| XM_017020138.1 | 11 | 2690 | XP_016875627.1 |
Target Gene Details
Entrez Gene ID: | 89894 |
Gene Name: | transmembrane protein 116 |
Gene Aliases: |
- |
Location: |
Chr.12:111894936-112013219 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM116 | XM_011538948.2 | XP_011537250.1 | ||
| XM_017020187.1 | XP_016875676.1 | |||
| AL833501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044606 | Chr.12:111885161 - 111989037 on Build GRCh38 | Gain |
|
| nsv1045488 | Chr.12:111883935 - 111922897 on Build GRCh38 | Gain |
|
| esv1010936 | Chr.12:111890837 - 111902032 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map