Assay Details
Target Gene Details
Entrez Gene ID: | 283431 |
Gene Name: | growth arrest specific 2 like 3 |
Gene Aliases: |
G2L3 |
Location: |
Chr.12:100573661-100628288 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS2L3 | NM_001303130.1 | NP_001290059.1 | ||
| NM_174942.2 | NP_777602.1 | |||
| XM_005268804.3 | XP_005268861.1 | |||
| XM_011538219.2 | XP_011536521.1 | |||
| XM_011538220.2 | 1 | 141 | XP_011536522.1 | |
| XM_017019206.1 | XP_016874695.1 | |||
| AK315189.1 | ||||
| BC043366.1 | AAH43366.1 | |||
| BP872524.1 |
More Information
Set Membership: |
|

Genomic Map