Assay Details
Target Gene Details
Entrez Gene ID: | 59341 |
Gene Name: | transient receptor potential cation channel subfamily V member 4 |
Gene Aliases: |
BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC |
Location: |
Chr.12:109783087-109833407 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
More Information
Additional Information:
For this assay, SNP(s) [rs140630874] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map