Assay Details
Target Gene Details
Entrez Gene ID: | 1337 |
Gene Name: | cytochrome c oxidase subunit 6A1 |
Gene Aliases: |
CMTRID, COX6A, COX6AL |
Location: |
Chr.12:120438090-120440742 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX6A1 | NM_004373.3 | NP_004364.2 | ||
| AK312009.1 | ||||
| BP378295.1 | ||||
| HE647860.1 | ||||
| X15341.1 | CAA33392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630923 | Chr.12:120439963 - 120450931 on Build GRCh38 | Gain |
|
| nsv509482 | Chr.12:120435117 - 120495113 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map