Assay Details
Target Gene Details
Entrez Gene ID: | 65244 |
Gene Name: | spermatogenesis associated serine rich 2 |
Gene Aliases: |
Nbla00526, P59SCR, SCR59, SPATA10 |
Location: |
Chr.12:49366905-49527441 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATS2 | NM_001293285.1 | NP_001280214.1 | ||
| NM_001293286.1 | NP_001280215.1 | |||
| NM_023071.3 | NP_075559.2 | |||
| NR_120663.1 | ||||
| XM_011538690.2 | XP_011536992.1 | |||
| XM_011538691.2 | XP_011536993.1 | |||
| XM_017019840.1 | XP_016875329.1 | |||
| AK023202.1 | BAB14460.1 | |||
| AK095582.1 | ||||
| AK292334.1 | ||||
| AL833614.1 | ||||
| BC048299.1 | AAH48299.1 | |||
| BX648916.1 | ||||
| CN421379.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832405 | Chr.12:49326496 - 49517079 on Build GRCh38 | Gain |
|
| dgv1497n100 | Chr.12:49284746 - 49438835 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map