Assay Details
Target Gene Details
Entrez Gene ID: | 8825 |
Gene Name: | lin-7 homolog A, crumbs cell polarity complex component |
Gene Aliases: |
LIN-7A, LIN7, MALS-1, TIP-33, VELI1 |
Location: |
Chr.12:80792520-80937915 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIN7A | NM_001324423.1 | 5 | 5850 | NP_001311352.1 |
| NM_004664.3 | 6 | 5969 | NP_004655.1 | |
| NR_136887.1 | 6 | 5927 | ||
| NR_136888.1 | 7 | 6046 | ||
| XM_011538928.2 | 5 | 5744 | XP_011537230.1 | |
| AK022997.1 | 1 | 2646 |
More Information
Set Membership: |
|

Genomic Map