Assay Details
Target Gene Details
Entrez Gene ID: | 3843 |
Gene Name: | importin 5 |
Gene Aliases: |
IMB3, KPNB3, Pse1, RANBP5, imp5 |
Location: |
Chr.13:97953641-98024296 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO5 | NM_002271.4 | NP_002262.3 | ||
| XM_005254052.3 | XP_005254109.1 | |||
| XM_011521087.2 | XP_011519389.1 | |||
| XM_011521088.2 | XP_011519390.1 | |||
| XM_011521089.2 | XP_011519391.1 | |||
| XM_017020562.1 | XP_016876051.1 | |||
| AL705630.1 | ||||
| BC045640.1 | AAH45640.1 | |||
| Y08890.1 | CAA70103.1 |
More Information
Set Membership: |
|

Genomic Map