Assay Details
Target Gene Details
Entrez Gene ID: | 51131 |
Gene Name: | PHD finger protein 11 |
Gene Aliases: |
APY, BCAP, IGEL, IGER, IGHER, NY-REN-34, NYREN34 |
Location: |
Chr.13:49495610-49528992 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF11 | NM_001040443.2 | NP_001035533.1 | ||
| NM_001040444.2 | NP_001035534.1 | |||
| NR_135322.1 | ||||
| NR_135323.1 | ||||
| AB011031.1 | BAA32101.1 | |||
| AF155105.1 | AAD42871.1 | |||
| AK026228.1 | ||||
| AK225986.1 | ||||
| AK293242.1 | ||||
| AL552215.3 | ||||
| BC017212.2 | AAH17212.2 | |||
| BX461254.2 |
More Information
Set Membership: |
|

Genomic Map