Assay Details
Target Gene Details
Entrez Gene ID: | 100190939 |
Gene Name: | TPT1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:45341345-45391483 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPT1-AS1 | NR_024458.1 | |||
| AF318337.1 | AAL55844.1 | |||
| AK124332.1 | BAC85835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631951 | Chr.13:45352364 - 45361056 on Build GRCh38 | Loss |
|
| nsv510579 | Chr.13:45305226 - 45424938 on Build GRCh38 | Deletion |
|
| esv3580625 | Chr.13:45348477 - 45359056 on Build GRCh38 | Loss |
|
| esv3403190 | Chr.13:45356317 - 45357715 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76895826] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map