Assay Details
Target Gene Details
Entrez Gene ID: | 51371 |
Gene Name: | proteasome maturation protein |
Gene Aliases: |
C13orf12, HSPC014, PNAS-110, UMP1 |
Location: |
Chr.13:28659004-28678957 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POMP | NM_015932.5 | NP_057016.1 | ||
| GU045555.1 | ||||
| GU045556.1 |
More Information
Set Membership: |
|

Genomic Map