Assay Details
Target Gene Details
Entrez Gene ID: | 51371 |
Gene Name: | proteasome maturation protein |
Gene Aliases: |
C13orf12, HSPC014, PNAS-110, UMP1 |
Location: |
Chr.13:28659004-28678957 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POMP | NM_015932.5 | NP_057016.1 | ||
| AF077200.1 | AAD26995.1 | |||
| AF125097.1 | AAD39914.1 | |||
| AF262975.1 | AAK58521.1 | |||
| AF275807.1 | AAG23819.1 | |||
| AK312118.1 | ||||
| BC003390.1 | AAH03390.1 | |||
| BC014334.1 | AAH14334.1 | |||
| EF535527.1 | ||||
| GU045555.1 | ||||
| GU045556.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75218575] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map