Assay Details
Target Gene Details
Entrez Gene ID: | 221150 |
Gene Name: | spindle and kinetochore associated complex subunit 3 |
Gene Aliases: |
C13orf3, RAMA1 |
Location: |
Chr.13:21153595-21176602 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKA3 | NM_001166017.1 | NP_001159489.1 | ||
| NM_145061.5 | NP_659498.4 | |||
| XM_005266288.3 | XP_005266345.1 | |||
| XM_011534994.1 | XP_011533296.1 | |||
| AF361358.1 | AAO13796.1 | |||
| AK090584.1 | BAC03484.1 | |||
| AK314864.1 | ||||
| BC048988.1 | AAH48988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3078n54 | Chr.13:21153703 - 21158036 on Build GRCh38 | Gain |
|
| nsv512307 | Chr.13:21154471 - 21157207 on Build GRCh38 | Loss |
|
| esv8341 | Chr.13:21156501 - 21157351 on Build GRCh38 | Loss |
|
| dgv924n106 | Chr.13:21155811 - 21157948 on Build GRCh38 | Deletion |
|
| esv2175517 | Chr.13:21156577 - 21157277 on Build GRCh38 | Deletion |
|
| nsv1037082 | Chr.13:20932196 - 21204957 on Build GRCh38 | Gain |
|
| esv1003302 | Chr.13:21156551 - 21157299 on Build GRCh38 | Deletion |
|
| nsv1149576 | Chr.13:21155868 - 21158050 on Build GRCh38 | Deletion |
|
| esv3631454 | Chr.13:21155131 - 21157989 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map