Assay Details
Target Gene Details
Entrez Gene ID: | 4093 |
Gene Name: | SMAD family member 9 |
Gene Aliases: |
MADH6, MADH9, PPH2, SMAD8, SMAD8/9, SMAD8A, SMAD8B |
Location: |
Chr.13:36844831-36920721 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD9 | NM_001127217.2 | 7 | 4881 | NP_001120689.1 |
| NM_005905.5 | 6 | 4770 | NP_005896.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522336 | Chr.13:36731504 - 37010701 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map