Assay Details
Target Gene Details
Entrez Gene ID: | 22862 |
Gene Name: | fibronectin type III domain containing 3A |
Gene Aliases: |
FNDC3, HUGO, bA203I16.1, bA203I16.5 |
Location: |
Chr.13:48975241-49209779 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNDC3A | NM_001079673.1 | NP_001073141.1 | ||
| NM_001278438.1 | NP_001265367.1 | |||
| NR_103528.1 | ||||
| XM_017020440.1 | XP_016875929.1 | |||
| XM_017020441.1 | XP_016875930.1 | |||
| XM_017020442.1 | XP_016875931.1 | |||
| XM_017020443.1 | XP_016875932.1 | |||
| AJ749706.1 | CAG44602.1 | |||
| AK056071.1 | ||||
| BC132812.1 | ||||
| BC136617.1 | ||||
| BC144301.1 | ||||
| BX640739.1 | CAE45852.1 | |||
| BX648141.1 | CAI45989.1 | |||
| BX648706.1 | ||||
| DB460317.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034 | Chr.13:48952625 - 48998591 on Build GRCh38 | Deletion |
|
| nsv455882 | Chr.13:48957415 - 49105021 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map