Assay Details
Target Gene Details
Entrez Gene ID: | 5095 |
Gene Name: | propionyl-CoA carboxylase alpha subunit |
Gene Aliases: |
- |
Location: |
Chr.13:100089015-100530437 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
Target Gene Details
Entrez Gene ID: | 100885777 |
Gene Name: | PCCA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:100464437-100481015 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCCA-AS1 | NR_047686.1 | |||
| AA430203.1 | ||||
| AA758845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv977273 | Chr.13:100469380 - 100471840 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map