Assay Details
Target Gene Details
Entrez Gene ID: | 10284 |
Gene Name: | Sin3A associated protein 18 |
Gene Aliases: |
2HOR0202, SAP18P |
Location: |
Chr.13:21140514-21149085 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAP18 | NM_005870.4 | NP_005861.2 | ||
| XM_011534868.2 | XP_011533170.1 | |||
| AF153608.1 | AAD41090.1 | |||
| AK311748.1 | ||||
| AY550970.1 | AAT52216.1 | |||
| BC030836.1 | AAH30836.1 | |||
| CR457035.1 | CAG33316.1 | |||
| DB171768.1 | ||||
| U78303.1 | AAF21220.1 | |||
| U96915.1 | AAC51322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037082 | Chr.13:20932196 - 21204957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map