Assay Details
Target Gene Details
Entrez Gene ID: | 7178 |
Gene Name: | tumor protein, translationally-controlled 1 |
Gene Aliases: |
HRF, TCTP, p02, p23 |
Location: |
Chr.13:45333471-45341284 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPT1 | NM_001286272.1 | 6 | 1415 | NP_001273201.1 |
| NM_001286273.1 | 5 | 1176 | NP_001273202.1 | |
| NM_003295.3 | 6 | 1250 | NP_003286.1 | |
| BC022436.1 | AAH22436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510579 | Chr.13:45305226 - 45424938 on Build GRCh38 | Deletion |
|
| nsv1046200 | Chr.13:45274249 - 45351310 on Build GRCh38 | Gain |
|
| nsv951078 | Chr.13:45329366 - 45340765 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map