Assay Details
Target Gene Details
Entrez Gene ID: | 7750 |
Gene Name: | zinc finger MYM-type containing 2 |
Gene Aliases: |
FIM, MYM, RAMP, SCLL, ZNF198 |
Location: |
Chr.13:19957412-20091845 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMYM2 | NM_001190964.2 | NP_001177893.1 | ||
| NM_001190965.2 | NP_001177894.1 | |||
| NM_003453.4 | NP_003444.1 | |||
| NM_197968.3 | NP_932072.1 | |||
| XM_005266520.3 | XP_005266577.1 | |||
| XM_017020729.1 | XP_016876218.1 | |||
| XM_017020731.1 | XP_016876220.1 | |||
| XM_017020732.1 | XP_016876221.1 | |||
| XM_017020733.1 | XP_016876222.1 | |||
| XM_017020734.1 | XP_016876223.1 | |||
| XM_017020735.1 | XP_016876224.1 | |||
| XM_017020736.1 | XP_016876225.1 | |||
| XM_017020737.1 | XP_016876226.1 | |||
| XM_017020738.1 | XP_016876227.1 | |||
| AJ224901.1 | CAA12204.1 | |||
| AK226118.1 | ||||
| AK289741.1 | ||||
| AL136621.1 | CAB66556.2 | |||
| BC036372.1 | AAH36372.1 | |||
| BI460760.1 | ||||
| BX643074.1 | ||||
| BX647944.1 | CAH56193.1 | |||
| Y13472.1 | CAA73875.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832557 | Chr.13:19910435 - 20081915 on Build GRCh38 | Gain |
|
| nsv1054226 | Chr.13:19511073 - 19989089 on Build GRCh38 | Gain |
|
| nsv561215 | Chr.13:19871303 - 20076371 on Build GRCh38 | Gain |
|
| esv2761244 | Chr.13:19389134 - 20043200 on Build GRCh38 | Gain |
|
| nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain |
|
| esv3892297 | Chr.13:19803308 - 20022425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map