Assay Details
Target Gene Details
Entrez Gene ID: | 160851 |
Gene Name: | diacylglycerol kinase eta |
Gene Aliases: |
DGKeta |
Location: |
Chr.13:42040036-42256584 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGKH | NM_001204504.2 | NP_001191433.1 | ||
| NM_152910.5 | NP_690874.2 | |||
| NM_178009.4 | NP_821077.1 | |||
| AB078967.1 | BAC66960.1 | |||
| AB078968.1 | BAC66961.1 | |||
| AK289546.1 | ||||
| DR002010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016 | Chr.13:42041861 - 42072289 on Build GRCh38 | Insertion |
|
| nsv832589 | Chr.13:42062545 - 42203497 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs536867463] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map