Assay Details
Target Gene Details
Entrez Gene ID: | 10910 |
Gene Name: | SGT1 homolog, MIS12 kinetochore complex assembly cochaperone |
Gene Aliases: |
SGT1 |
Location: |
Chr.13:52652691-52700756 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUGT1 | XM_011534891.2 | 14 | 5210 | XP_011533193.1 |
| XM_011534893.2 | 13 | 4928 | XP_011533195.1 | |
| XM_017020365.1 | 14 | 5100 | XP_016875854.1 | |
| XM_017020366.1 | 14 | 4975 | XP_016875855.1 | |
| XM_017020367.1 | 14 | 5009 | XP_016875856.1 | |
| XM_017020368.1 | 14 | 5021 | XP_016875857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044949 | Chr.13:52497005 - 52715291 on Build GRCh38 | Gain |
|
| dgv1653n100 | Chr.13:52564997 - 52728939 on Build GRCh38 | Gain |
|
| nsv820041 | Chr.13:52487203 - 52706453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map