Assay Details
Target Gene Details
Entrez Gene ID: | 79596 |
Gene Name: | ring finger protein 219 |
Gene Aliases: |
C13orf7 |
Location: |
Chr.13:78614286-78659179 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF219 | NM_024546.3 | NP_078822.3 | ||
| XM_011535225.1 | XP_011533527.1 | |||
| XM_017020743.1 | XP_016876232.1 | |||
| AK098640.1 | ||||
| AL136651.1 | CAB66586.3 | |||
| BC136764.1 | ||||
| BC136765.1 | ||||
| BI550796.1 | ||||
| CR936752.1 | ||||
| CR936754.1 | ||||
| CR936764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2421781 | Chr.13:78628966 - 78633507 on Build GRCh38 | Deletion |
|
| dgv340n67 | Chr.13:78628719 - 78634373 on Build GRCh38 | Loss |
|
| esv2667717 | Chr.13:78628856 - 78634363 on Build GRCh38 | Deletion |
|
| nsv517259 | Chr.13:78631277 - 78633453 on Build GRCh38 | Loss |
|
| esv3632802 | Chr.13:78628868 - 78634405 on Build GRCh38 | Loss |
|
| dgv339n67 | Chr.13:78560929 - 78651758 on Build GRCh38 | Gain |
|
| nsv1098 | Chr.13:78622118 - 78655578 on Build GRCh38 | Insertion |
|
| nsv1160140 | Chr.13:78628317 - 78635700 on Build GRCh38 | Deletion |
|
| esv2747700 | Chr.13:78628870 - 78634370 on Build GRCh38 | Deletion |
|
| dgv234n27 | Chr.13:78566708 - 78642283 on Build GRCh38 | Loss |
|
| nsv514712 | Chr.13:78629216 - 78633440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map