Assay Details
Target Gene Details
Entrez Gene ID: | 10562 |
Gene Name: | olfactomedin 4 |
Gene Aliases: |
GC1, GW112, OLM4, OlfD, UNQ362, bA209J19.1, hGC-1, hOLfD |
Location: |
Chr.13:53028741-53052061 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLFM4 | NM_006418.4 | NP_006409.3 | ||
| AF097021.1 | AAC72970.1 | |||
| AK000683.1 | ||||
| AK300947.1 | ||||
| AY358567.1 | AAQ88930.1 | |||
| BC047740.1 | AAH47740.1 | |||
| BC117329.1 | ||||
| DA928098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832613 | Chr.13:52950184 - 53113053 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map