Assay Details
Target Gene Details
Entrez Gene ID: | 51676 |
Gene Name: | ankyrin repeat and SOCS box containing 2 |
Gene Aliases: |
ASB-2 |
Location: |
Chr.14:93934153-93976796 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB2 | NM_001202429.1 | NP_001189358.1 | ||
| NM_016150.4 | NP_057234.2 | |||
| XM_005267758.3 | XP_005267815.1 | |||
| XM_011536834.2 | XP_011535136.1 | |||
| XM_011536835.2 | XP_011535137.1 | |||
| XM_017021369.1 | XP_016876858.1 | |||
| AB056723.1 | BAB64532.1 | |||
| AB488462.1 | ||||
| AF159164.1 | AAD45345.1 | |||
| AJ251238.1 | CAC17765.1 | |||
| AK002049.1 | ||||
| AK057078.1 | ||||
| AK093271.1 | ||||
| AK303686.1 | ||||
| AK315628.1 | ||||
| AL137735.1 | CAB70899.1 | |||
| BC032354.1 | AAH32354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832861 | Chr.14:93768697 - 93936069 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map