Assay Details
Target Gene Details
Entrez Gene ID: | 51562 |
Gene Name: | MAP3K12 binding inhibitory protein 1 |
Gene Aliases: |
- |
Location: |
Chr.14:36298558-36320676 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBIP | NM_001144891.1 | NP_001138363.1 | ||
| NM_001308110.1 | NP_001295039.1 | |||
| NM_016586.2 | NP_057670.2 | |||
| XM_005267754.3 | XP_005267811.1 | |||
| XM_005267756.4 | XP_005267813.1 | |||
| XM_017021367.1 | XP_016876856.1 | |||
| AB038523.1 | BAA94083.1 | |||
| AF208857.1 | AAF64271.1 | |||
| AK293473.1 | ||||
| AK315290.1 | ||||
| BC005197.1 | AAH05197.1 | |||
| BC016821.1 | AAH16821.1 | |||
| BX248287.1 | CAD62615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054427 | Chr.14:36311537 - 36350725 on Build GRCh38 | Gain |
|
| nsv832771 | Chr.14:36301801 - 36494836 on Build GRCh38 | Loss |
|
| nsv1040487 | Chr.14:36301607 - 36336415 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map