Assay Details
Target Gene Details
Entrez Gene ID: | 4331 |
Gene Name: | MNAT1, CDK activating kinase assembly factor |
Gene Aliases: |
CAP35, MAT1, RNF66, TFB3 |
Location: |
Chr.14:60734735-60969881 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MNAT1 | NM_001177963.1 | NP_001171434.1 | ||
| NM_002431.3 | NP_002422.1 | |||
| XM_005267687.2 | XP_005267744.1 | |||
| XM_005267688.2 | XP_005267745.1 | |||
| XM_017021333.1 | XP_016876822.1 | |||
| AA053721.1 | ||||
| AK308701.1 | ||||
| BC000820.1 | AAH00820.1 | |||
| CD701743.1 | ||||
| CR450336.1 | CAG29332.1 | |||
| KU178191.1 | ||||
| KU178192.1 | ||||
| U61835.1 | AAB05248.1 | |||
| X87843.1 | CAA61112.1 | |||
| X92669.1 | CAA63356.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037387 | Chr.14:60742756 - 61066784 on Build GRCh38 | Gain |
|
| nsv564880 | Chr.14:60397865 - 60916029 on Build GRCh38 | Gain |
|
| nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139789698] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map