Assay Details
Target Gene Details
Entrez Gene ID: | 283578 |
Gene Name: | transmembrane p24 trafficking protein family member 8 |
Gene Aliases: |
FAM15B |
Location: |
Chr.14:77335021-77377384 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED8 | NM_213601.1 | NP_998766.1 | ||
| XM_017021223.1 | XP_016876712.1 | |||
| XM_017021225.1 | XP_016876714.1 | |||
| AK095650.1 | ||||
| AY598329.1 | AAT06740.1 | |||
| BC101510.1 | AAI01511.1 |
More Information
Set Membership: |
|

Genomic Map