Assay Details
Target Gene Details
Entrez Gene ID: | 317761 |
Gene Name: | chromosome 14 open reading frame 39 |
Gene Aliases: |
Six6os1 |
Location: |
Chr.14:60435954-60499773 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C14orf39 | NM_174978.2 | NP_777638.2 | ||
| XM_011536690.2 | XP_011534992.1 | |||
| XM_011536703.2 | XP_011535005.1 | |||
| XM_011536704.1 | XP_011535006.1 | |||
| XM_017021247.1 | XP_016876736.1 | |||
| XM_017021248.1 | XP_016876737.1 | |||
| XM_017021249.1 | XP_016876738.1 | |||
| XM_017021250.1 | XP_016876739.1 | |||
| XM_017021251.1 | XP_016876740.1 | |||
| AK098187.1 | BAC05253.1 | |||
| AL832219.1 | ||||
| BC125070.1 | ||||
| DB447488.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892552 | Chr.14:60416097 - 60478741 on Build GRCh38 | Loss |
|
| nsv564880 | Chr.14:60397865 - 60916029 on Build GRCh38 | Gain |
|
| nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map