Assay Details
Target Gene Details
Entrez Gene ID: | 26030 |
Gene Name: | pleckstrin homology and RhoGEF domain containing G3 |
Gene Aliases: |
ARHGEF43, KIAA0599 |
Location: |
Chr.14:64704129-64744346 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHG3 | NM_001308147.1 | NP_001295076.1 | ||
| XM_011536624.2 | XP_011534926.1 | |||
| XM_011536625.2 | XP_011534927.1 | |||
| XM_011536628.2 | XP_011534930.1 | |||
| XM_011536629.2 | XP_011534931.1 | |||
| XM_017021154.1 | XP_016876643.1 | |||
| BC129952.1 | ||||
| BC129953.1 | ||||
| BX248779.1 | CAD66586.1 | |||
| CN284264.1 | ||||
| HY057390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035833 | Chr.14:64709653 - 64779463 on Build GRCh38 | Gain |
|
| nsv1309 | Chr.14:64681609 - 64714932 on Build GRCh38 | Insertion |
|
| nsv1049097 | Chr.14:64561035 - 64735300 on Build GRCh38 | Gain |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map