Assay Details
Target Gene Details
Entrez Gene ID: | 55334 |
Gene Name: | solute carrier family 39 member 9 |
Gene Aliases: |
ZIP-9, ZIP9 |
Location: |
Chr.14:69398379-69462390 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A9 | NM_001252148.1 | NP_001239077.1 | ||
| NM_001252150.1 | NP_001239079.1 | |||
| NM_001252151.1 | NP_001239080.1 | |||
| NM_001252152.1 | NP_001239081.1 | |||
| NM_018375.4 | NP_060845.2 | |||
| XM_005267834.1 | XP_005267891.1 | |||
| XM_017021426.1 | XP_016876915.1 | |||
| AK002136.1 | BAA92100.1 | |||
| AK222547.1 | BAD96267.1 | |||
| AK293222.1 | ||||
| AK310285.1 | ||||
| AK311104.1 | ||||
| AK316381.1 | ||||
| AY358687.1 | AAQ89050.1 | |||
| AY780789.1 | AAV98359.1 | |||
| BC047682.1 | AAH47682.1 | |||
| BC064383.1 | AAH64383.1 | |||
| BP299863.1 | ||||
| BX643103.1 | ||||
| FJ423549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3634836 | Chr.14:69388471 - 69421903 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map