Assay Details
Target Gene Details
Entrez Gene ID: | 9321 |
Gene Name: | thyroid hormone receptor interactor 11 |
Gene Aliases: |
ACG1A, CEV14, GMAP-210, GMAP210, TRIP-11, TRIP230 |
Location: |
Chr.14:91965991-92040140 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIP11 | NM_001321851.1 | NP_001308780.1 | ||
| NM_004239.4 | NP_004230.2 | |||
| XM_017021787.1 | XP_016877276.1 | |||
| XM_017021788.1 | XP_016877277.1 | |||
| AF007217.1 | AAD09135.1 | |||
| AF011368.1 | AAB84386.1 | |||
| BC146845.1 | ||||
| BX641024.1 | CAE46015.1 | |||
| L40380.1 | AAC41730.1 | |||
| Y12490.1 | CAA73095.1 |
More Information
Set Membership: |
|

Genomic Map