Assay Details
Target Gene Details
Entrez Gene ID: | 79696 |
Gene Name: | zinc finger C2HC-type containing 1C |
Gene Aliases: |
C14orf140, FAM164C |
Location: |
Chr.14:75069577-75079987 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC2HC1C | NM_001042430.2 | 3 | 2599 | NP_001035895.1 |
| NM_024643.3 | 3 | 3114 | NP_078919.2 | |
| NR_110315.1 | 2 | 1757 |
More Information
Set Membership: |
|

Genomic Map