Assay Details
Target Gene Details
Entrez Gene ID: | 55860 |
Gene Name: | actin-related protein 10 homolog |
Gene Aliases: |
ACTR11, Arp10, Arp11, HARP11 |
Location: |
Chr.14:58200115-58235636 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR10 | NM_018477.2 | NP_060947.1 | ||
| XM_011536960.1 | XP_011535262.1 | |||
| XM_011536961.1 | XP_011535263.1 | |||
| XM_011536962.2 | XP_011535264.1 | |||
| XM_011536963.2 | XP_011535265.1 | |||
| AF220190.1 | AAF67655.1 | |||
| AK000544.1 | BAA91243.1 | |||
| AK022534.1 | BAB14083.1 | |||
| AK222624.1 | BAD96344.1 | |||
| AK222702.1 | BAD96422.1 | |||
| AK315537.1 | ||||
| BC005208.1 | ||||
| BC011997.1 | AAH11997.1 | |||
| BX161491.1 | CAD61940.1 | |||
| BX248282.1 | CAD62610.1 |
More Information
Set Membership: |
|

Genomic Map