Assay Details
Target Gene Details
Entrez Gene ID: | 78990 |
Gene Name: | OTU deubiquitinase, ubiquitin aldehyde binding 2 |
Gene Aliases: |
C14orf137, OTB2, OTU2 |
Location: |
Chr.14:94016083-94048933 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTUB2 | NM_023112.3 | NP_075601.1 | ||
| XM_005268055.3 | XP_005268112.1 | |||
| AK025569.1 | BAB15172.1 | |||
| BC009615.1 | AAH09615.1 | |||
| BC068058.1 | AAH68058.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34067 | Chr.14:94024355 - 94160851 on Build GRCh38 | Loss |
|
| nsv565620 | Chr.14:93990701 - 94031622 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74074370] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map