Assay Details
Target Gene Details
Entrez Gene ID: | 5583 |
Gene Name: | protein kinase C eta |
Gene Aliases: |
PKC-L, PKCL, PRKCL, nPKC-eta |
Location: |
Chr.14:61280953-61550980 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCH | XM_017021458.1 | XP_016876947.1 | ||
| AK296158.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76412640] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map