Assay Details
Target Gene Details
Entrez Gene ID: | 254028 |
Gene Name: | uncharacterized LOC254028 |
Gene Aliases: |
- |
Location: |
Chr.14:20588224-20607221 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC254028 | NR_122043.1 | |||
| AK093210.1 | ||||
| BI830942.1 | ||||
| DB034207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|

Genomic Map